Motic BA310 LED Compound Microscope | Avantor
Motic BA310 LED Compound Microscope
BA310 digital LED compound microscope
Catalog # CA76635-266
Supplier:   Motic
Fournisseur #:  1100401200721
Motic BA310 LED Compound Microscope
Catalog # CA76635-266
Supplier:  Motic
Motic BA310 LED Compound Microscope
Motic BA310 LED Compound Microscope
BA310 digital LED compound microscope
Price
$3,911.54
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
Product Details & Documents
The BA310 Elite model introduces superior improvements both in optical and mechanical performance. It is designed for the rigors of daily routine work in the demanding applications of universities, clinics, laboratories, and many other life sciences or medical applications requiring quality optical performance.
  • Widefield high eyepoint eyepiece N-WF10X/20 mm with diopter
  • Adjustment on both eyepieces, with rubber eyecup (paired)
  • Reversed quintuple revolving nosepiece
  • Coaxial coarse and fine focusing adjustment with tension adjustment
  • Built in low position coaxial mechanical stage (right-hand control)
  • Focusable Abbe condenser N.A. 0.9 with iris diaphragm and slot
The ELITE model's addition of a new rackless stage concept without prominent gear rack allows an even more convenient use of the x/y movement. For full freedom of illumination options, the new model has a complete and easy interchangeability of its 6 V/30 W halogen bulb with LED modules.
Ordering information: Contact your Avantor representative to learn more about this and other motic solutions. All motic products come with a one-year electrical and five-year mechanical warranty.
Spécifications
  • Désignation:
    BA310 digital LED compound microscope