About this item
The BA310E model introduces superior improvements both in optical and mechanical performance. The ELITE model's addition of a new rackless stage concept without prominent gear rack allows an even more convenient use of the x/y movement. For full freedom of illumination options, the new model has a complete and easy interchangeability of its 6 V/30 W Halogen bulb with LED modules.
- BA310E
- Trinocular head Siedentopf type 30° inclined, 360° swiveling, 360° rotating (light split 100:0/20:80)
- Widefield high eyepoint eyepiece N-WF10×/20 mm
- With diopter adjustment on both eyepieces and rubber eyecups
- Reversed quintuple revolving nosepiece
- 3WLED module for BA210E/BA310E (6000K)
- Moticam BMH4000X
- 1920×1080 HDMI monitor with attachable microscopy camera with 4K (3840×2160) resolution in SD card
- 4K through HDMI
- 20fps at 3840×2160 through USB connection
- 12 V power adapter, CS ring, Motic 4-dot calibration slide, dust cap, manual, 12 mm focusable lens, 30 and 38 mm eyepiece adapters, macro tube, macro calibration card
- Motic Images Plus 3.1 for Windows/macOS
- Moticam BTI10
- 10" LCD Android tablet with 1280x800 screen resolution
- Connection Wi-Fi (2.4 and 5 GHz), Ethernet
- WiFi resolution 4 MP (2592×1520 pixels) captured resolution
- WiFi output (802.1 a/b/g/n/ac supporting both hot-spot mode or using existing WiFi network)
- Preloaded optimized MotiConnect app
The Moticam BMH4000X is a camera that was designed for ease of use and the ability to output fast live images at 4K (3840×2160) pixels in full HD. Unlike other cameras, the Moticam BMH4000X can be connected to a computer or through HDMI. Simply plug the camera through the HDMI port and display images with the use of the onboard software and a USB mouse through the built-in 11.6" LCD screen, or connect your camera through USB to a Windows, Mac or Linux computer.
The Moticam BTI10 unlike other Moticams is combined with a 4.0 MP and a 10" Android tablet that will deliver fast live images up to 30 fps on any trinocular microscope with the right C-mount adapter. With the convenience of a detachable tablet and WiFi image data transfer, the flexibility to capture images and bring your images anywhere has never been so easy.