Motic BA210E LED Compound Microscopes | Avantor
Motic BA210E LED Compound Microscopes
Supplier:   VWR International
Motic BA210E LED Compound Microscopes
Supplier:  VWR International
Certificates
Motic BA210E LED Compound Microscopes
Motic BA210E LED Compound Microscopes
5 Options Available Below
Product Details & Documents
The BA210Elite model introduces superior improvements both in optical and mechanical performance. Meant to be used in Educational life sciences, Medical and a variety of Biological applications, the BA210E is designed for ease of use and longevity due to Motic's complete understanding of the educational market.
  • Widefield high eyepoint eyepiece N-WF10×/20 mm with diopter
  • Adjustment on both eyepieces, with rubber eyecup (paired)
  • Reversed quadruple revolving nosepiece
  • Coaxial coarse and fine focusing adjustment
  • Built in low position coaxial mechanical stage (right-hand control)
  • Focusable Abbe condenser N.A. 1.25 with iris diaphragm and slot
The BA210E model introduces student-proof features both in optical and mechanical aspects. The new rackless stage concept without prominent gear rack allows even more convenient use of the x/y movement, while increasing user safety in teaching situations.
For full freedom of illumination options, the new model BA210E has an easy interchangeability of its 6 V/30 W Halogen bulb with LED modules of different colour temperatures (4500K, 6000K). Extended life-time of LEDs here is combined with cost saving aspects. Light consuming contrast methods like Phase contrast, Polarization or Dark field can easily performed.
Ordering information: Contact your Avantor representative to learn more about this and other motic solutions. All motic products come with a one-year electrical and five-year mechanical warranty.
Delivery information: The BA210E Phase package option (76163-614) includes upright compound microscope, trinocular head, widefield high eyepoint 10× eyepieces, CCIS EC Plan achromat/phase objectives (PL4×, PL100× S-Oil, Ph10X), rackless stage, NA 1.25 abbe condenser, phase slider, phase centering telescope, 3WLED module.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • BA210 Digital LED compound microscope
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,082.96
    $3,082.96
    Each
       $3,082.96
    La quantité minimale de commande est de 1
  • BA210E Binocular LED compound microscope
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,788.78
    $1,788.78
    Each
       $1,788.78
    La quantité minimale de commande est de 1
  • BA210E Phase package
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,412.15
    $1,412.15
    Each
       $1,412.15
  • BA210E Phase package
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,906.08
    $2,906.08
    Each
       $2,906.08
    La quantité minimale de commande est de 1
  • BA210E Trinocular LED compound microscope
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,058.15
    $2,058.15
    Each
       $2,058.15
    La quantité minimale de commande est de 1