Millennia™ 2000 Adhesion Slides
 :   StatLab
Millennia™ 2000 Adhesion Slides
 :  StatLab
Millennia™ 2000 Adhesion Slides
Millennia™ 2000 Adhesion Slides
Millennia™ 2000 slides feature a hydrophilic surface with strong adhesion properties.
  • Hydrophilic slide designed for IHC, special stains, frozen sections, and routine histology
  • Thermal transfer printer compatible
  • Available with 45 degree clipped corners only
Part of the StatLab Millennia Adhesion Microscope Slide family. Manufactured in Japan.
 : Slides are packaged 100 per box.
Product Family Options
  • Color
    Épaisseur
    L×l
  • Blue
    1 mm
    75×25 mm
    Box of 100
    Box of 100 - 69,05 $
    69,05 $
    Each
    Each - 69,05 $
    69,05 $
    Box of 100
       69,05 $
    Each
       69,05 $
     : M2000B
  • Green
    1 mm
    75×25 mm
    Box of 100
    Box of 100 - 69,05 $
    69,05 $
    Each
    Each - 69,05 $
    69,05 $
    Box of 100
       69,05 $
    Each
       69,05 $
     : M2000G
  • Pink
    1 mm
    75×25 mm
    Box of 100
    Box of 100 - 69,05 $
    69,05 $
    Each
    Each - 69,05 $
    69,05 $
    Box of 100
       69,05 $
    Each
       69,05 $
     : M2000P
  • White
    1 mm
    75×25 mm
    Box of 100
    Box of 100 - 69,05 $
    69,05 $
    Each
    Each - 69,05 $
    69,05 $
    Box of 100
       69,05 $
    Each
       69,05 $
     : M2000W
  • Yellow
    1 mm
    75×25 mm
    Box of 100
    Box of 100 - 69,05 $
    69,05 $
    Each
    Each - 69,05 $
    69,05 $
    Box of 100
       69,05 $
    Each
       69,05 $
     : M2000Y