Millennia™ Command Adhesion Slides
White, 1 mm, 75×25 mm
Catalog # 76548-148
Supplier: StatLab
Fournisseur #: MCOMM/W/90
Jump to:
Product Details & Documents
A universal adhesion slide for H&E, IHC, and special stains.
- Hybrid slide suitable for IHC, H&E, and special staining
- Optimized coating for strong adhesion to reduce tissue loss and provide consistency of staining
- Exhibits minimal background staining during all types of staining
- Hydrophilic, 90 degree corners
- Compatible with thermal transfer printers
Part of the StatLab Millennia Adhesion Microscope Slide family. Manufactured in Japan.
Packaging: Slides are packaged 100 per box.
Spécifications
- Color:White
- L×l:75×25 mm
- Épaisseur:1 mm