Essentials Disposable Long-Handled Scoops, HDPE, Antylia Scientific
30 cm long handle makes it easier to reach into deep containers.
- Manufactured and packed in a Class 100,000 cleanroom to ensure cleanliness and reduce cross contamination
- Long handle makes it more visible when left in the container
- Made from high-quality HDPE which conforms to FDA, EC, and EU regulations
- BSE/TSE free
- Non-Sterile
Use these disposable long-handled spoons to dispense powders and granules. Manufactured in a cleanroom, they are individually packaged and can be used straight out of the bag without cleaning. Long handles ensure hands are away from the materials being measured.
: Supplier Certificate of Conformity is included with product shipment.
Frequently Bought Together