Bucket, Vileda
Longish Bucket for use with CE Bucket Trolley or CE Pre-Prepared Trolley, Grey, 25 L, 22 x 10 x 11"
Catalog # 89413-093
Supplier: Vileda Professional - FHP
Fournisseur #: 131662
Jump to:
Product Details & Documents
Polypropylene bucket with stainless steel handle. For use on the CE Bucket Trolley or the CE Pre-Prepared Trolley.
- Liquid measurement lines molded inside of bucket
- Color coding clips included to allow differentiation in Colors of buckets (red, blue, green, yellow)
- Autoclavable
Spécifications
- Capacité:25 L
- Color:Grey
- Désignation:Longish Bucket for use with CE Bucket Trolley or CE Pre-Prepared Trolley
- Dimensions:22 x 10 x 11"