Coriolis® Air Sampler Accessories, Bertin Instruments
Coriolis® Air Sampler Accessories, Bertin Instruments
Supplier:   BERTIN CORP
Coriolis® Air Sampler Accessories, Bertin Instruments
Supplier:  BERTIN CORP
Certificates
Coriolis® Air Sampler Accessories, Bertin Instruments
Coriolis® Air Sampler Accessories, Bertin Instruments
4 Options Available Below
Product Details & Documents
Coriolis® air sampler consumables are part of the innovative dry cyclonic technology, which aspirates airborne particles and centrifuges them in a cone.
  • Single use consumables
  • Ensure optimal collection efficiency for Coriolis® air samplers
The separation of the airborne particles from the air flow is due to the air flow rate, the air intake geometry, the design of the cones, and the collection liquid (surfactant in low concentration). The collected particles or microorganisms can then be recovered by rinsing the consumables with an appropriate buffer solution, so the sample is compatible with multiple downstream analysis options (NGS, qPCR, culture).
Product Family Options
Product Information
  • Désignation
    Matière
    Availability
    Price
  • Sterile cone
    Polycarbonate
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $1,449.63
    $1,449.63
    Pack of 50
       $1,449.63
    Minimum order quantity is 1
    Supplier Number: S001112-CORM0-A.0
  • Sterile cone assembled with cap
    Polycarbonate
    Log in for availability
    Log in for availability
    Pack of 5
    Pack of 5 - $221.17
    $221.17
    Pack of 5
       $221.17
    Minimum order quantity is 1
    Supplier Number: S001109-CORM0-A.0
  • Sterile cone assembled with cap
    Polycarbonate
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $911.41
    $911.41
    Pack of 50
       $911.41
    Minimum order quantity is 1
    Supplier Number: S001111-CORM0-A.0
  • Sterile cone with air intake
    Polycarbonate
    Discontinued
    Supplier Number: -
  • Sterile cone, liquid dose
    Polycarbonate
    Discontinued
    Supplier Number: -
  • Sterile cone, liquid dose
    Polycarbonate
    Discontinued
    Supplier Number: -
  • Sterile cone, liquid dose
    Polycarbonate
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $640.98
    $640.98
    Pack of 50
       $640.98
    Minimum order quantity is 1
    Supplier Number: S001113-CORM0-A.0