Coriolis® COMPACT Air Sampler, Bertin Instruments
Coriolis® COMPACT portable dry air sampler, 50 L/min
Catalog # 76636-420
Supplier:   BERTIN CORP
 :  P002055-CORC0-A.0
Coriolis® COMPACT Air Sampler, Bertin Instruments
Catalog # 76636-420
Supplier:  BERTIN CORP
Coriolis® COMPACT Air Sampler, Bertin Instruments
Coriolis® COMPACT Air Sampler, Bertin Instruments
Coriolis® COMPACT portable dry air sampler, 50 L/min
Price
$16,616.97
Log in for availability
Each
Product Details & Documents
Portable dry air sampler for bio contamination control.
  • Concentrates airborne particles using a dry cyclonic technique
  • Compatible with culture and molecular biology standard methods (PCR, qPCR, or NGS)
  • Easy decontamination (hydrogen peroxide)
  • Autonomy on battery of 8 hours
  • Light and compact for indoor and outdoor use
Coriolis® COMPACT is a portable biological air sampler using a dry cyclonic technique, for indoor or outdoor air bio-contamination monitoring. Its innovative dry cyclonic technology aspirates the particles with an airflow of 50 L/min and centrifuges them in a cone. The collected particles or microorganisms (bacteria, viruses, moulds) can then be recovered by rinsing with an appropriate buffer that can be analysed afterwards using microbiological techniques.
Sample types include virus, bacteria, moulds, pollens, and spores.
Suitable for environmental and pollution research, pharmaceutical, food and veterinary industries, biomedical and health environments.
 : Plastic cones are used with a screw cap to facilitate the transport and preservation of samples. Sterile and non sterile cones and caps are available (ready to use doses).
Specifications
  • Débit:
    50 L/min
  • Désignation:
    Coriolis® COMPACT portable dry air sampler
  • Cat. No.:
    76636-420
  • Particle size:
    0.5 - 10 µm
  • Running time:
    1 min - 8 h
  • W×D×H:
    135×130×225 mm
  • Weight:
    1.2 kg (with battery)
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Coriolis® COMPACT cones and air intake
    Coriolis® COMPACT cones and air intake
    76636-422
    Log in for availability
    Pack of 10 - $225.41