miniPCR® P51™ Molecular Fluorescence Viewer
Catalogue # 470348-600
Fournisseur: AMPLYUS LLC
Spécifications
- Description:miniPCR® P51™ molecular fluorescence viewer
- Cat. no.:470348-600
Spécifications
À propos de cet article
Visualize DNA, RNA, and proteins at work right in the palm of your hand.
- Tube holder design allows for pipetting while tubes are in the viewer
- Views 8 tubes at once
- Power source either through 9V battery or USB-C
- Includes orange filter for performing BioBits and P51 Molecular Glow Labs
- Bonus yellow filter for enhanced viewing of yellow and green fluorescence
With P51™, you can detect and measure molecules like DNA, RNA and proteins in a simple, visual way. Scientists use fluorescence every day to detect and quantify DNA, RNA, and proteins, but this often requires sophisticated and complex equipment. With P51™, we have packaged the power to visualize molecules into a compact, cheap, handheld format that’s perfect for any setting.
Through hands-on investigations, students will explore the building blocks of life by visualizing them in the P51™ Fluorescence Viewer. Simply place your samples inside the P51™ and turn on the safe blue light – and see your tubes light up!