Ward's® Tiger Eye
Supplier:   Avantor
Ward's® Tiger Eye
Supplier:  Avantor
Certificates
Ward's® Tiger Eye
Ward's® Tiger Eye
2 Options Available Below
Product Details & Documents
Unpolished metamorphic rock with silky lusture, red-brown colour, and clear parallel intergrown quartz crystals.
  • Collected in Africa
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
  • Matching thin section available
Student specimens - Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand specimens - Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
All specimen dimensions are considered an average size range.
Product Family Options
Specimens
Informations sur le produit
  • Désignation
    Dimensions
    Specimen Dimensions
    Availability
    Price
  • Tiger eye, student specimens
    1×1" - 1×2"
    1×1 - 1×1"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $30.15
    $30.15
    Each
       $30.15
  • Tiger eye, hand specimens
    2×3" - 3×4"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $30.00
    $30.00
    Each
       $30.00
Storage
Informations sur le produit
  • Catalog #
    Désignation
    Availability
    Price
  • Storage box
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2.35
    $2.35
    Each
       $2.35