Bioquell SeQure Bio-Decontamination System | Avantor
Bioquell SeQure Bio-Decontamination System
Bioquell SeQure Bio-Decontamination System
Catalog # 76635-466
Supplier:   Eco Lab
Fournisseur #:  VWRBQSQRE
Bioquell SeQure Bio-Decontamination System
Catalog # 76635-466
Supplier:  Eco Lab
Bioquell SeQure Bio-Decontamination System
Bioquell SeQure Bio-Decontamination System
Bioquell SeQure Bio-Decontamination System
Price
Renseignez-vous sur le prix
Log in pour la disponibilité
Each
Product Details & Documents
The Bioquell SeQure is a fixed and compact wall-mounted bio-decontamination system. The technology utilizes Bioquell Hydrogen Peroxide Sterilant to kill microorganisms on exposed surfaces in spaces <150m3 . It offers a small footprint with fast installation and customized validated cycles performed by Ecolab validation experts on-site.
  • Fixed hydrogen peroxide vapor generator for chambers or rooms
  • Create validated cycles for 6-log sporicidal kill on exposed surfaces utilizing Bioquell Hydrogen Peroxide Sterilant (EPA Registration Number 72372-1-86703)
  • Fast to install and validate (<1 week)
  • Bottle module with peroxide traceability and wastage minimization through the two-bottle module system
  • Ideal for pass-throughs, material airlocks, small enclosures and rooms less than 150m3 (5297 ft3)
  • Designed with RFID label technology for traceability and auditing
  • 21 CFR Part 11 audit trail compliance package
  • Integration testing for BMS and more complex systems
  • Cycle recording (thermal printer and solutions for real time cycle data, including electronic records in pdf format)
  • Supports GMP compliance
The Bioquell SeQure is a fixed and compact wall-mounted bio-decontamination system. The technology utilizes Bioquell Hydrogen Peroxide Sterilant (EPA Registration Number 72372-1-86703) to kill microorganisms on exposed surfaces in spaces <150m3. It offers a small footprint with fast installation and customized validated cycles performed by Ecolab validation experts on-site.
The wall-mounted system is ideal for pass-throughs, material airlocks, and more. The Bioquell SeQure system features automatic cycle prediction technology for fast validation. Whether installed in a brand-new facility or retrofitted into an existing workspace, the Bioquell SeQure can be installed and validated in facilities in less than one week. The slim design and digital, user-friendly interface make the Bioquell SeQure ideal for use in many rooms and chambers. Its minimal footprint is enabled by wall-mounting the components of the system inside or outside of the workspace.
Spécifications
  • Désignation:
    Bioquell SeQure Bio-Decontamination System