Bioquell Room CI | Avantor
Bioquell Room CI
Room Chemical Indicator (Room CI), 100
Catalog # 76635-444
Supplier:   Eco Lab
Fournisseur #:  10250284
Bioquell Room CI
Catalog # 76635-444
Supplier:  Eco Lab
Bioquell Room CI
Bioquell Room CI
Room Chemical Indicator (Room CI), 100
Price
$1,086.10
Log in pour la disponibilité
Case of 100
La quantité minimale de commande est de 1
Product Details & Documents
Bioquell offers different indicators to help you to verify bio-decontamination cycle completion and effectiveness. Chemical indicators (CI) can help you to verify a bio-decontamination cycle has achieved the expected vapor distribution and exposure. The Bioquell Room CI is for rooms greater than 10m3
  • rapid-read chemical indicators for Ecolab Bioquell hydrogen peroxide vapor bio-decontamination
  • easy-to- read color changing results for rooms and enclosed areas greater than 10m3
  • Quickly confirm expected vapor distribution and exposure performance.
Confirm that a Bioquell cycle has achieved the expected vapor distribution and exposure performance with these easy-to-read Bioquell Room CIs (Chemical Indicators). A chemical strip reacts to a Bioquell cycle and changes color to correlate with the cycle's actual vapor exposure. For use in rooms and enclosed areas 10m3 and larger.
Spécifications
  • Désignation:
    Room Chemical Indicator (Room CI)
  • Taille:
    100