Bioquell Isolator CI
Isolator Chemical Indicator (Isolator CI), 100, Isolator CI
Catalog # 76635-440
Supplier: Eco Lab
Fournisseur #: 10250075
Jump to:
- L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Return Policy
Product Details & Documents
Bioquell offers different indicators to help you to verify bio-decontamination cycle completion and effectiveness. Chemical indicators (CI) can help you to verify a bio-decontamination cycle has achieved the expected vapor distribution and exposure. The Bioquell Isolator CI is for enclosures less than 10m3.
- rapid-read chemical indicators for Ecolab Bioquell hydrogen peroxide vapor bio-decontamination for enclosed areas smaller than 10m3
- Ideal for isolators, BSCs, filling lines, or small areas such as pass-throughs and MALs
- Quickly confirm expected vapor distribution and exposure performance.
The Bioquell Isolator CIs are rapid-read chemical indicators for enclosed areas smaller than 10m^3. Use when bio-decontaminating isolators, BSCs, filling lines, or small areas such as pass-throughs and MALs, to quickly confirm expected vapor distribution and exposure performance.
Spécifications
- Désignation:Isolator Chemical Indicator (Isolator CI)
- Dimensions:Isolator CI
- Taille:100