Sterile SatPax® MicroSeal®-VP Pre-wetted Sealed Edge Wipes, 100% Polyester Knit
Supplier: Berkshire
New Product
Certificates
About this item
Sterile SatPax® MicroSeal®-VP is an ultrasonically sealed edge cleanroom laundered wipe recommended for ISO Class 3 and above environments. This wipe is composed of 100% continuous filament polyester knit fabric.
- Pre-wetted with 70% USP Grade IPA
- Ultrasonically sealed edge for reduced fiber contamination
- Laundered and packaged in Berkshire's ISO Class 4 cleanroom with lot number on each pack
- Lightweight, high absorbency material design reduces actual cost in use compared to heavier weight products
- Critically low particles, fibers, ions, and extractables
Sterile SatPax® MicroSeal®-VP wipes are pre-wetted with a consistent concentration of IPA and DI water and a 100% IPA option. This combination of properties provides Sterile SatPax® MicroSeal ®-VP with the highest cleanliness, abrasion resistance, and chemical compatibility required for cleaning critical environments. The pre-wetted format offers a cost-effective and easy-to-use solution versus traditional bulk handling of solvents and maintenance of squirt bottles.
Certifications: ISO Class 3 - 5, Cleanroom Class 1 - 100. EU Grade B/C/D.
Packaging: Wipes are packaged 30 sheets/pack, 24 packs/case (J-fold).
Frequently Bought Together