PUREZERO* HG5 White Non Sterile Nitrile Gloves
Supplier: O&M HALYARD CANADA, INC CA
New Product
Certificates
About this item
Halyard PUREZERO* HG5 White Nitrile Cleanroom Gloves are designed for semiconductor, pharmaceutical and medical device manufacturing applications. They are recommended for use in ISO Class 5 or higher and grade B/C/D cleanrooms.
- Ambidextrous design
- Textured fingertips, with tacky grip
- AQL 1.0
- Latex-free, powder-free and silicone-free
- Low dermatitis potential
- Static dissipative in use
- Max particle count: 1500
PUREZERO* HG5 White Nitrile Cleanroom Gloves are clean processed (washed repeatedly in deionized water) to ensure consistent control of low particles and extractables. Due to an accelerator-free formulation, there is a reduced risk of allergies and skin irritation associated with accelerator chemicals in these nitrile gloves.
Certifications: Gloves meet USP 800 guidelines.
Specifications
- Catalog No:
- 76635-372
- 76635-374
- 76635-380
- 76635-378
- 76635-376
- 77575-304
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Material:
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Finish:
- Tacky grip
- Tacky grip
- Tacky grip
- Tacky grip
- Tacky grip
- Tacky grip
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile