ContactUsLinkComponent
Presto™ Laser Histology Microscope Slide Printer, Azer Scientific
Catalogue # 76632-834
Fournisseur:  VWR International
undefined
Presto™ Laser Histology Microscope Slide Printer, Azer Scientific
Catalogue # 76632-834
Fournisseur:  VWR International
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Spécifications
  • Description:
    Laser histology microscope slide printer; 100-slide cartridge capacity, up to 35-slide output tray capacity
  • Dimensions:
    46L×20W×34H cm (18.1×7.9×13.4")
  • Cat. no.:
    76632-834
  • Weight:
    19 kg (42 lbs.)
Spécifications
À propos de cet article
Azer Presto™ Microscope Slide Printer allows for quick, permanent and efficient printing on microscope slides. UV laser technology eliminates the need of expensive printer consumables and maintenance.
  • Integrate to LIS/HIS system
  • High resolution 500×500 dpi
  • High speed; as fast as three seconds for on-demand printing
  • Input cartridge holds up to 100 slides and output holds up to 35 slides
  • Continuous and on-demanding printing model
  • Barcode, QR code, logo, alphanumeric, graphic printing
  • ≤65dB work noise
Azer Scientific's Presto™ microscope slide printer is an ultraviolet laser model that can integrate into any laboratory or hospital information system. Our UV laser technology creates a permanent labeling resistant to degradation. Implementing the new Azer Scientific Presto™ slide printer aids in a time management workflow that you will not want to resist.
Ordering information: Two year warranty.
Delivery: Includes two pre-installed templates. Contact prior to shipping for data fields required.
Caution: Do not attempt to open during operation.