Capacitors
Capacitors
Compact electric storage for circuits.
  • 25 V
  • Radial design
  • Connects easily to bred boards
Capacitors are useful when you need to store a charge and release it in a circuit. These consist of two parallel plate capacitors that are arranged in a rolled-up design to minimize its volume.
Product Family Options
  • Désignation
  • Capacitor, 25 V, 100 µF
    Each
    Each - 0,85 $
    0,85 $
    Pack of 10
    Pack of 10 - 8,45 $
    8,45 $
    Each
       0,85 $
    Pack of 10
       8,45 $
     : 25V100UFPK10
  • Capacitor, 25 V, 220 µF
    Each
    Each - 0,90 $
    0,90 $
    Pack of 10
    Pack of 10 - 9,25 $
    9,25 $
    Each
       0,90 $
    Pack of 10
       9,25 $
     : 25V220UFPK10
  • Capacitor, 25 V, 330 µF
    Each
    Each - 0,90 $
    0,90 $
    Pack of 10
    Pack of 10 - 9,25 $
    9,25 $
    Each
       0,90 $
    Pack of 10
       9,25 $
     : 25V330UFPK10