Zener Diode | Avantor
Zener Diode
Zener diode
Catalog # 470347-906
Fournisseur #:  DIZEN500MWPK10
Zener Diode
Catalog # 470347-906
Zener Diode
Zener Diode
Zener diode
Liquidation
0,25 $
Vente
Valable jusqu'au 31/12/2026
Log in pour la disponibilité
Each Pack of 10
Product Details & Documents
Control the current direction in circuits.
  • 2 pin connectors
  • Fits all standard bred boards
Zener diodes are a unique electrical component that allows the current to flow in a “reverse direction”. A diode only allows a current to flow in one direction and prevents an electrical charge being reversed. A Zener diode does allow the current flow to be reversed, but only once the voltage equals the specific voltage rating.
Spécifications
  • Désignation:
    Zener diode
  • Cat. No.:
    470347-906