REUZ™ Stainless Steel Pharma Scoops
Reusable stainless steel scoops for powders and granulated materials.
- 304 stainless steel
- Smooth, polished surface
- Easy to clean
- Large handle
- Available in sizes from 10 to 2500 ml
Manufactured with 304 stainless steel, REUZ™ ('re-use') Pharma Scoops have polished surfaces and rounded edges. The precision corrosion-resistant stainless construction includes large handles and smooth, large radius interior surfaces for easy cleaning and protection against bacteria deposits. Includes large rims near the handle to protect against spills when the scoop is tilted up. Intended for handling powders and granules in laboratory and production settings.
REUZ™ products promote sustainability in all laboratory environments.
: Sold individually.
Frequently Bought Together