Eisco Glass Volumetric Flasks, Class B
Supplier: Wards
Certificates
About this item
Class B volumetric flasks without stoppers are made from borosilicate 3.3 glass and feature a graduation mark, blue enamel markings, and beaded rims.
- Made of high quality, heavy duty, borosilicate 3.3 glass for excellent durability and chemical resistance
- Class B tolerances conform with DIN ISO 1042 standards for accuracy
- Easy-to-read blue enamel graduations and specifications for contrast against clear liquids
- Fire-polished beaded rims are chip and crack resistant
Calibrated to contain.
Eisco labs borosilicate glass has the following heat tolerances:
- Strain point 515 °C
- Annealing point 565 °C
- Softening point 820 °C
Frequently Bought Together