Eisco Plastic Volumetric Flasks | Avantor
Eisco Plastic Volumetric Flasks
Supplier:   Wards
Eisco Plastic Volumetric Flasks
Supplier:  Wards
Certificates
Eisco Plastic Volumetric Flasks
Eisco Plastic Volumetric Flasks
2 Options Available Below
Product Details & Documents
Polypropylene volumetric flasks with single graduation line and screw caps.
  • Made from high-quality polypropylene
  • Highly chemical resistant, microwavable, and autoclavable at 121 °C
  • Tight-fitting screw cap to prevent leakage of contents
Basic volumetric flasks are made of heavy duty polypropylene plastic with a high heat tolerance, temperature range of −20 to +135 °C, and good to excellent chemical resistance. Autoclavable at 121 °C for easy cleaning and sterilization. Good contact clarity. Fitted with screw cap to prevent leakage. Marked with a single graduation line and provided with a screw cap.
Product Family Options
Informations sur le produit
  • Capacité
    Availability
    Price
  • 100 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 15,10 $
    15,10 $
    Each
       15,10 $
    Supplier Number: CH0452C
  • 250 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 17,05 $
    17,05 $
    Each
       17,05 $
    Supplier Number: CH0452D