About this item
GenScript offers a wide selection of Protein molecular weight standards for SDS-PAGE and Western blot, including prestained, unstained, peptide, Western-viewable, and other standards.
- Comprehensive selection: a variety of standards available for SDS-PAGE and Western blotting
- Broad molecular weight range: low, middle and broad MW range standards suitable for all practical needs
- Easy identification: clean, sharp protein bands for accurate MW estimation and assessment of transfer efficiency
The Pre-stained Protein Standards consist of colored bands for easy identification and allow you to monitor the progress of an electrophoretic run. The Unstained Protein Standards provide bands that are unmodified by the presence of dye, for accurate molecular weight estimation of SDS-PAGE. Available in convenient package sizes, all standards have undergone stringent QC to assure consistent performance to provide the sharper bands in the broader molelular weight range available.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.