K5 Monochrome sCMOS Camera, Leica Microsystems
CA76485-962
- Model:K5
- Resolution:4.2 MP
- Sensor Type:sCMOS
- Sensor size:18.8 mm
- Frame rate:40 fps (full resolution)
- Supported OS:Windows 10
- Pixel size:6.5×6.5 μm
- Pixels (W×H):2048×2048 mm
- Dynamic range:21400:1 up to 87 dB
- Interfaces:USB 3.1
- Power consumption:4.5 W
- Sensor:sCMOS
- Software:LAS X
- Weight:420 g
- Cat. no.:CA76485-962
For routine fluorescence imaging.
- Bring the flexibility and power of sCMOS technology into your lab and image the details of your experiment
- Efficiently segment crucial, real-time events
- Improve signal-to-background of your THUNDER imager
Image a wide variety of sample types with the flexibility and power of high 4.2 megapixel resolution and fast 40 fps imaging brought to you by the K5. Recommended for live imaging and lower signal-to-noise fluorescence imaging, especially when paired with a Leica THUNDER imager.