ContactUsLinkComponent
Adjustable Boss Head Clamp
  470343-240
undefined
Adjustable Boss Head Clamp
  470343-240
 :  4-ACH40-10
  • Description:
    Boss head clamp
  • Cat. no.:
    470343-240
These boss head clamp holders are adjustable up to 16 mm.
  • Accepts rods up to 5/8" in diameter
  • Made with cast-iron body and nickel-plated thumb-screws