PYREX® Stopcocks Keys, Corning | Avantor
PYREX® Stopcocks Keys, Corning
   Options durables disponibles
Supplier:   Corning
PYREX® Stopcocks Keys, Corning
Supplier:  Corning
   Options durables disponibles
Certificates
PYREX® Stopcocks Keys, Corning
4 Options Available Below
Product Details & Documents
These are replacement PTFE stopcock plug assemblies with various diameter bores for PYREX® straight bore PTFE stopcocks.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 29610-634
  • 29610-640
  • 29610-642
  • 29610-638
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Ø trou
    For
    Availability
    Price
  • Replacement PTFE product standard straight bore stopcock plug assembly, O-ring size A
    2 mm
    Straight bore PTFE stopcocks
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $162.00
    $162.00
    Case of 1
       $162.00
    Supplier Number: 7681-2
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    29610-634
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • Replacement PTFE product standard straight bore stopcock plug assembly, O-ring size B
    6 mm
    Straight bore PTFE stopcocks
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $318.00
    $318.00
    Case of 1
       $318.00
    Supplier Number: 7681-6
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    29610-640
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • Replacement PTFE product standard straight bore stopcock plug assembly, O-ring size C
    8 mm
    Straight bore PTFE stopcocks
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $356.00
    $356.00
    Case of 1
       $356.00
    Supplier Number: 7681-8
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    29610-642
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • Replacement PTFE product standard straight bore stopcock plug assembly, O-ring size B
    4 mm
    Straight bore PTFE stopcocks
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $231.00
    $231.00
    Case of 1
       $231.00
    Supplier Number: 7681-4
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    29610-638
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact