Undercarriages for angled tubs, Remco
Fournisseur: Remco Products
Certificats
À propos de cet article
These galvanized steel undercarriages feature caster wheels that are impervious to meat fat and bearings equipped with grease fittings.
- Replacement casters and wheels are available
- Regular model includes a built-in stop that allows the tub to be tipped and emptied
- Low undercarriage is ideal when mobility is required, but tipping is not necessary
These tub accessories are commonly used for meat, seafood, poultry, commercial fishing, confectionery, bakery, and grain applications.
For use with Remco tubs 6901*.
Frequently Bought Together