AssureTouch™ Power Gloves
À propos de cet article
Gloves feature extended cuffs to provide extra protection for wrists and forearms.
- Extended cuffs provide optimal coverage
- Textured fingertips ensure a secure grip, even when wet
- Chemo tested and fentanyl safe
These gloves provide strong puncture, tear, and chemical resistance without compromising dexterity or tactility.
Packaging: Gloves are packaged 100 gloves/box, 10 boxes/case (1000 gloves/case).
8 Options Available Below
Product Details & Documents
Gloves feature extended cuffs to provide extra protection for wrists and forearms.
- Extended cuffs provide optimal coverage
- Textured fingertips ensure a secure grip, even when wet
- Chemo tested and fentanyl safe
These gloves provide strong puncture, tear, and chemical resistance without compromising dexterity or tactility.
Packaging: Gloves are packaged 100 gloves/box, 10 boxes/case (1000 gloves/case).
Specifications for Product Family
Spécifications
- Catalog No:
- 76579-736
- 76579-738
- 76579-742
- 76579-744
- CA76579-737
- CA76579-739
- CA76579-741
- CA76579-743
- CA76579-745
- CA76579-736
- CA76579-738
- CA76579-740
- CA76579-742
- CA76579-744
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Finish:
- Finger textured
- Finger textured
- Finger textured
- Finger textured
- Finger textured
- Finger textured
- Finger textured
- Finger textured
- Finger textured
- Finger textured
- Finger textured
- Finger textured
- Finger textured
- Finger textured
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Extended
- Extended
- Extended
- Extended
- Extended
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Cuff length:
- 12"
- 12"
- 12"
- 12"
- 0.00276"
- 0.00276"
- 0.00276"
- 0.00276"
- 0.00276"
- 12"
- 12"
- 12"
- 12"
- 12"
Spécifications
Product Family Options
Informations sur le produit
- Taille du gantGlove ColorLongueurÉpaisseurAvailabilityPrice
- XSBlue220 mm4 mil.Discontinued-Supplier Number: 9999-ASpécifications:
More
Specifications
Cat. No.76579-736Ambidextrous/Hand-specificAmbidextrousDisposable / reusableDisposableLatex/Latex-freeLatex-freePowdered/Powder-freePowder-freeFinishFinger texturedCuff styleBeadedCuff length12" - SBlue220 mm4 mil.Case of 1,000$235.42Each$235.42Supplier Number: 9999-BSpécifications:
More
Specifications
Cat. No.76579-738Ambidextrous/Hand-specificAmbidextrousDisposable / reusableDisposableLatex/Latex-freeLatex-freePowdered/Powder-freePowder-freeFinishFinger texturedCuff styleBeadedCuff length12" - LBlue230 mm4 mil.Case of 1,000$235.42Each$235.42Supplier Number: 9999-DSpécifications:
More
Specifications
Cat. No.76579-742Ambidextrous/Hand-specificAmbidextrousDisposable / reusableDisposableLatex/Latex-freeLatex-freePowdered/Powder-freePowder-freeFinishFinger texturedCuff styleBeadedCuff length12" - XLBlue230 mm4 mil.Case of 1,000$235.42Each$235.42Supplier Number: 9999-ESpécifications:
More
Specifications
Cat. No.76579-744Ambidextrous/Hand-specificAmbidextrousDisposable / reusableDisposableLatex/Latex-freeLatex-freePowdered/Powder-freePowder-freeFinishFinger texturedCuff styleBeadedCuff length12" - XSBlue220 mm3.2 mil.Box of 100$18.05Case of 1000$180.50Supplier Number: 9999-ASpécifications:
More
Specifications
Cat. No.CA76579-737Ambidextrous/Hand-specificAmbidextrousDisposable / reusableDisposableLatex/Latex-freeLatex-freePowdered/Powder-freePowder-freeFinishFinger texturedCuff styleExtendedCuff length0.00276" - SBlue220 mm3.2 mil.Box of 100$18.05Case of 1000$180.50Supplier Number: 9999-BSpécifications:
More
Specifications
Cat. No.CA76579-739Ambidextrous/Hand-specificAmbidextrousDisposable / reusableDisposableLatex/Latex-freeLatex-freePowdered/Powder-freePowder-freeFinishFinger texturedCuff styleExtendedCuff length0.00276" - MBlue230 mm3.2 mil.Box of 100$18.05Case of 1000$180.50Supplier Number: 9999-CSpécifications:
More
Specifications
Cat. No.CA76579-741Ambidextrous/Hand-specificAmbidextrousDisposable / reusableDisposableLatex/Latex-freeLatex-freePowdered/Powder-freePowder-freeFinishFinger texturedCuff styleExtendedCuff length0.00276" - LBlue230 mm3.2 mil.Box of 100$18.05Case of 1000$180.50Supplier Number: 9999-DSpécifications:
More
Specifications
Cat. No.CA76579-743Ambidextrous/Hand-specificAmbidextrousDisposable / reusableDisposableLatex/Latex-freeLatex-freePowdered/Powder-freePowder-freeFinishFinger texturedCuff styleExtendedCuff length0.00276" - XLBlue230 mm3.2 mil.Box of 100$18.05Case of 1000$180.50Supplier Number: 9999-ESpécifications:
More
Specifications
Cat. No.CA76579-745Ambidextrous/Hand-specificAmbidextrousDisposable / reusableDisposableLatex/Latex-freeLatex-freePowdered/Powder-freePowder-freeFinishFinger texturedCuff styleExtendedCuff length0.00276" - XSBlue220 mm4 mil.Discontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CA76579-736Ambidextrous/Hand-specificAmbidextrousDisposable / reusableDisposableLatex/Latex-freeLatex-freePowdered/Powder-freePowder-freeFinishFinger texturedCuff styleBeadedCuff length12" - SBlue220 mm4 mil.Discontinued-Supplier Number: 9999-BSpécifications:
More
Specifications
Cat. No.CA76579-738Ambidextrous/Hand-specificAmbidextrousDisposable / reusableDisposableLatex/Latex-freeLatex-freePowdered/Powder-freePowder-freeFinishFinger texturedCuff styleBeadedCuff length12" - MBlue230 mm4 mil.Discontinued-Supplier Number: 9999-CSpécifications:
More
Specifications
Cat. No.CA76579-740Ambidextrous/Hand-specificAmbidextrousDisposable / reusableDisposableLatex/Latex-freeLatex-freePowdered/Powder-freePowder-freeFinishFinger texturedCuff styleBeadedCuff length12" - LBlue230 mm4 mil.Discontinued-Supplier Number: 9999-DSpécifications:
More
Specifications
Cat. No.CA76579-742Ambidextrous/Hand-specificAmbidextrousDisposable / reusableDisposableLatex/Latex-freeLatex-freePowdered/Powder-freePowder-freeFinishFinger texturedCuff styleBeadedCuff length12" - XLBlue230 mm4 mil.Discontinued-Supplier Number: 9999-ESpécifications:
More
Specifications
Cat. No.CA76579-744Ambidextrous/Hand-specificAmbidextrousDisposable / reusableDisposableLatex/Latex-freeLatex-freePowdered/Powder-freePowder-freeFinishFinger texturedCuff styleBeadedCuff length12"