OsmoTECH® PRO Multi-Sample Micro-Osmometer, Advanced Instruments | Avantor
OsmoTECH® PRO Multi-Sample Micro-Osmometer, Advanced Instruments
Multi-sample osmometer
Catalog # 76579-778
Fournisseur #:  OSMOTECH_PRO
OsmoTECH® PRO Multi-Sample Micro-Osmometer, Advanced Instruments
Catalog # 76579-778
OsmoTECH® PRO Multi-Sample Micro-Osmometer, Advanced Instruments
OsmoTECH® PRO Multi-Sample Micro-Osmometer, Advanced Instruments
Multi-sample osmometer
Price
Renseignez-vous sur le prix
Log in pour la disponibilité
Each
Product Details & Documents
Optimize and ensure the quality and consistency of buffers, media, product, excipients, and process parameters to maximize biologic yield, quality, and purity.
  • Multi-sample osmometer
  • Gold standard freezing point technology
  • Fully compliant with 21 CFR part 11, GMP, and EU Annex 11 with e-Signature
  • Meets Pharmacopeia osmolality testing guideline
Designed to meet the high sample throughput, regulatory compliance (including 21 CFR part 11), and data management needs of busy bioprocessing and biomanufacturing labs, the OsmoTECH PRO Multi-Sample Micro-Osmometer combines ease-of-use with accurate and precise osmolality measurements for reliable and reproducible results. Optimize and ensure the quality and consistency of buffers, media, product, excipients, and process parameters to maximize biologic yield, quality and purity.
Informations sur les accessoires: OsmoTECH® PRO Multi-Sample Micro-Osmometer IQ/OQ Service (sold separately): Eliminate delays with our IQ/OQ. Our comprehensive services ensure no workflow interruption so you can focus on your work. Our experts developed this comprehensive qualification package to comply with USP <1058> Analytical Instrument Qualification and USP <785> Osmolality and Osmolarity Testing. Don’t go it alone. Regulated environments require documented proof that your manufacturing process consistently produces quality product. Performing qualification internally is costly, time-consuming, and problematic. Our expert qualification services ensure fast and reliable commissioning of your instrument. Eliminate delays with our IQ/OQ. Our comprehensive services ensure no workflow interruption so you can focus on your work.
Spécifications
  • Désignation:
    Multi-sample osmometer
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Multi-sample osmometer convenience kit
    Multi-sample osmometer convenience kit
    76579-784
    Log in pour la disponibilité
    Each - $9,921.35
Informations sur le produit
  • Catalog #
    Details
    Availability
    Price
  • Multi-sample osmometer IQ/OQ service
    Multi-sample osmometer IQ/OQ service
    76579-770
    Installation and operational qualification: At instrument purchase, verifies and documents instrument delivery, installation and configuration as per manufacturer’s standards; Requalification: Ensures performance of instrument within operating ranges as per specifications
    Log in pour la disponibilité
    Each - $27,391.21