PYREX® Cylindrical Jars with Handles, Corning
Fournisseur: Corning
Certificats
À propos de cet article
These PYREX® jars are mold-blown with substantial walls for mechanical strength and have recessed finger grips for convenience in handling.
- Open end is ground flat
These jars can also be used as animal jars.
Caution: Do not use with heat, vacuum, or pressure applications.
Frequently Bought Together