Corning® External Thread Cryogenic Vials, Corning
   Options durables disponibles
Supplier:   Corning
Corning® External Thread Cryogenic Vials, Corning
Supplier:  Corning
   Options durables disponibles
Certificates
Corning® External Thread Cryogenic Vials, Corning
5 Options Available Below
Product Details & Documents
These vials are manufactured from polypropylene to withstand temperatures down to –196 °C.
  • Black graduations with large marking spot
  • Vials have a silicone washer for a secure seal
  • 2 ml self-standing vials are available in five cap colors.
  • Vials may be color coded with inserts
  • Free foam rack with each case
  • RNase-/DNase-free
  • Sterile
  • Nonpyrogenic
Self-standing vials have a special base design allowing them to be locked into cryogenic rack and tray (Cat. No. 431131) for single-handed manipulation.
 : Do not use cryogenic vials for storage in the liquid phase of liquid nitrogen. Only store vials in the vapor phase above the liquefied gas. Always use appropriate safety equipment when removing vials from cryogenic storage.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 66021-972
  • 66021-976
  • 66021-974
  • 66021-978
  • 66021-980
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
Spécifications
Product Family Options
Informations sur le produit
  • Capacité
    Bottom Style
    Avec bouchon
    Availability
    Price
  • 1.20 ml
    Conical bottom, self-standing
    Oui
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 500
    Case of 500 - $481.46
    $481.46
    Prix promotionnel pour les nouveaux clients
    Case of 500
       $481.46
    Supplier Number: 430658
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    66021-972
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 2.00 ml
    Round bottom
    Oui
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 500
    Case of 500 - $457.30
    $457.30
    Prix promotionnel pour les nouveaux clients
    Case of 500
       $457.30
    Supplier Number: 430661
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    66021-976
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 2.00 ml
    Round bottom, self-standing
    Oui
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 500
    Case of 500 - $700.70
    $700.70
    Prix promotionnel pour les nouveaux clients
    Case of 500
       $700.70
    Supplier Number: 430659
    Spécifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    66021-974
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
      Low Manufacturing Impact
  • 4.00 ml
    Round bottom, self-standing
    Oui
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 500
    Case of 500 - $573.48
    $573.48
    Prix promotionnel pour les nouveaux clients
    Case of 500
       $573.48
    Supplier Number: 430662
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    66021-978
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 5.00 ml
    Round bottom, self-standing
    Oui
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 500
    Case of 500 - $746.30
    $746.30
    Prix promotionnel pour les nouveaux clients
    Case of 500
       $746.30
    Supplier Number: 430663
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    66021-980
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact
Cap inserts
Informations sur le produit
  • Catalog #
    Désignation
    Color
    Availability
    Price
  • Color-Coded Cap Inserts
    Yellow
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 500
    Case of 500 - $95.20
    $95.20
    Case of 500
       $95.20
Informations sur le produit
  • Catalog #
    Désignation
    Color
    Availability
    Price
  • Color-Coded Cap Inserts
    Red
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 500
    Case of 500 - $95.20
    $95.20
    Case of 500
       $95.20
Informations sur le produit
  • Catalog #
    Désignation
    Color
    Availability
    Price
  • Color-Coded Cap Inserts
    Green
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 500
    Case of 500 - $95.20
    $95.20
    Case of 500
       $95.20
Informations sur le produit
  • Catalog #
    Désignation
    Color
    Availability
    Price
  • Color-Coded Cap Inserts
    White
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 500
    Case of 500 - $95.20
    $95.20
    Case of 500
       $95.20
Informations sur le produit
  • Catalog #
    Désignation
    Color
    Availability
    Price
  • Color-Coded Cap Inserts
    Blue
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 500
    Case of 500 - $95.20
    $95.20
    Case of 500
       $95.20
Informations sur le produit
  • Catalog #
    Désignation
    Color
    Availability
    Price
  • Color-Coded Cap Inserts
    Assorted
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 500
    Case of 500 - $92.65
    $92.65
    Case of 500
       $92.65