Corning® Internal Thread Cryogenic Vials, Corning
Corning® Internal Thread Cryogenic Vials, Corning
   Sustainable Options Available
Supplier:   Corning
Corning® Internal Thread Cryogenic Vials, Corning
Supplier:  Corning
   Sustainable Options Available
Certificates
Corning® Internal Thread Cryogenic Vials, Corning
14 Options Available Below
Product Details & Documents
These vials are manufactured from polypropylene to withstand temperatures down to –196 °C.
  • Black graduations with large marking spot
  • Vials have a silicone washer for a secure seal
  • 2 ml self-standing vials are available in five cap colors.
  • Vials may be color coded with inserts
  • Free foam rack with each case
  • RNase-/DNase-free
  • Sterile
  • Nonpyrogenic
Self-standing vials have a special base design allowing them to be locked into cryogenic rack and tray (Cat. No. 431131) for single-handed manipulation.
Caution: Do not use cryogenic vials for storage in the liquid phase of liquid nitrogen. Only store vials in the vapor phase above the liquefied gas. Always use appropriate safety equipment when removing vials from cryogenic storage.
Specifications for Product Family
Specifications
  • Catalog No:
  • 66021-940
  • 66021-944
  • 66021-946
  • 66021-950
  • 89089-764
  • 66021-942
  • 66021-948
  • 66021-970
  • 30617-556
  • 30617-558
  • 30617-560
  • 30617-562
  • 30617-564
  • 30617-566
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
Specifications
Product Family Options
Product Information
  • Capacité
    Bottom Style
    Avec bouchon
    Cap Color
    Availability
    Price
  • 1.20 ml
    Conical bottom, self-standing
    Oui
    Orange
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $457.30
    $457.30
    New Customer Promotional Price
    Case of 500
       $457.30
    Supplier Number: 430487
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    66021-940
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
      Low Manufacturing Impact
  • 2.00 ml
    Round bottom
    Oui
    Orange
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $650.25
    $650.25
    New Customer Promotional Price
    Case of 500
       $650.25
    Supplier Number: 430489
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    66021-944
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 4.00 ml
    Round bottom
    Oui
    Orange
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $639.20
    $639.20
    New Customer Promotional Price
    Case of 500
       $639.20
    Supplier Number: 430490
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    66021-946
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 5.00 ml
    Round bottom
    Oui
    Orange
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $746.30
    $746.30
    New Customer Promotional Price
    Case of 500
       $746.30
    Supplier Number: 430492
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    66021-950
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 2.00 ml
    Round bottom, self-standing
    Oui
    Orange
    Log in for availability
    Log in for availability
    Case of 250
    Case of 250 - $279.48
    $279.48
    New Customer Promotional Price
    Case of 250
       $279.48
    Supplier Number: 431386
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    89089-764
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 2.00 ml
    Round bottom, self-standing
    Oui
    Orange
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $543.00
    $543.00
    New Customer Promotional Price
    Case of 500
       $543.00
    Supplier Number: 430488
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    66021-942
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 4.00 ml
    Round bottom, self-standing
    Oui
    Orange
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $652.81
    $652.81
    New Customer Promotional Price
    Case of 500
       $652.81
    Supplier Number: 430491
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    66021-948
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 5.00 ml
    Round bottom, self-standing
    Oui
    Orange
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $746.30
    $746.30
    New Customer Promotional Price
    Case of 500
       $746.30
    Supplier Number: 430656
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    66021-970
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 2.00 ml
    Round bottom, self-standing
    Oui
    Assorted
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $573.75
    $573.75
    New Customer Promotional Price
    Case of 500
       $573.75
    Supplier Number: 431416
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    30617-556
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 2.00 ml
    Round bottom, self-standing
    Oui
    Yellow
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $521.05
    $521.05
    New Customer Promotional Price
    Case of 500
       $521.05
    Supplier Number: 431417
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    30617-558
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 2.00 ml
    Round bottom, self-standing
    Oui
    Blue
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $521.05
    $521.05
    New Customer Promotional Price
    Case of 500
       $521.05
    Supplier Number: 431418
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    30617-560
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 2.00 ml
    Round bottom, self-standing
    Oui
    Green
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $521.05
    $521.05
    New Customer Promotional Price
    Case of 500
       $521.05
    Supplier Number: 431419
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    30617-562
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 2.00 ml
    Round bottom, self-standing
    Oui
    Red
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $521.05
    $521.05
    New Customer Promotional Price
    Case of 500
       $521.05
    Supplier Number: 431420
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    30617-564
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 2.00 ml
    Round bottom, self-standing
    Oui
    White
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $521.05
    $521.05
    New Customer Promotional Price
    Case of 500
       $521.05
    Supplier Number: 431421
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    30617-566
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact
Cap inserts
Product Information
  • Catalog #
    Désignation
    Color
    Availability
    Price
  • Color-Coded Cap Inserts
    White
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $95.20
    $95.20
    Case of 500
       $95.20
Product Information
  • Catalog #
    Désignation
    Color
    Availability
    Price
  • Color-Coded Cap Inserts
    Blue
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $95.20
    $95.20
    Case of 500
       $95.20
Product Information
  • Catalog #
    Désignation
    Color
    Availability
    Price
  • Color-Coded Cap Inserts
    Red
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $95.20
    $95.20
    Case of 500
       $95.20
Product Information
  • Catalog #
    Désignation
    Color
    Availability
    Price
  • Color-Coded Cap Inserts
    Green
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $95.20
    $95.20
    Case of 500
       $95.20
Product Information
  • Catalog #
    Désignation
    Color
    Availability
    Price
  • Color-Coded Cap Inserts
    Yellow
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $95.20
    $95.20
    Case of 500
       $95.20
Product Information
  • Catalog #
    Désignation
    Color
    Availability
    Price
  • Color-Coded Cap Inserts
    Assorted
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $92.65
    $92.65
    Case of 500
       $92.65