Falcon® Feeder Tray, 24-Well, Corning
CA-BD351186
: Corning
- Description:Additional Feeder Tray with Lid for 24-Well Systems
- Cat. no.:CA-BD351186
- Supplier No.:351186
Large volume media feeder tray reduces the need for frequent media changes. Feeder tray allows all 24 wells to be bathed in the same medium for better uniformity across the plate.
For use with BD BioCoat* HTS Caco-2 assay systems.