Eisco Plastic Conical Flasks with Screw Cap
Eisco Plastic Conical Flasks with Screw Cap
Supplier:   Wards
Eisco Plastic Conical Flasks with Screw Cap
Supplier:  Wards
Certificates
Eisco Plastic Conical Flasks with Screw Cap
Eisco Plastic Conical Flasks with Screw Cap
3 Options Available Below
Product Details & Documents
Polypropylene conical flasks with screw caps and no graduations.
  • Made from high-quality polypropylene
  • Highly chemical resistant, microwavable and autoclavable at 121 °C
  • Tight-fitting screw caps for transportation and storage of contents
  • Wide neck for easy filling and cleaning
  • Translucent body makes it easy to see liquid level in the flask
Conical flasks with a wide neck and a screw cap. The natural translucent color makes it easy to see any liquid level in the bottle. Made from easy-to-clean high-quality polypropylene. Highly chemical resistant, microwavable and autoclavable at 121 °C.
Product Family Options
Product Information
  • Capacité
    Color
    Availability
    Price
  • 500 ml
    Translucent
    Log in for availability
    Log in for availability
    Each
    Each - $16.05
    $16.05
    Each
       $16.05
    Supplier Number: CH0431C
  • 100 ml
    Translucent
    Log in for availability
    Log in for availability
    Each
    Each - $7.10
    $7.10
    Each
       $7.10
    Supplier Number: CH0431A
  • 250 ml
    Translucent
    Log in for availability
    Log in for availability
    Each
    Each - $8.90
    $8.90
    Each
       $8.90
    Supplier Number: CH0431B