About this item
VWR® Taq Plus is an optimized format of Taq DNA polymerase master mix and, therefore, is a good alternative to Taq DNA polymerase and Taq DNA polymerase master mix.
- Ready to use 2X master mix for greater convenience
- Significantly reduced set-up time
- Increased specificity compared to Taq DNA polymerase master mix
- Reduced risk of contamination
- Processes up to 4 kb
Ensures increased specificity and improved PCR performance, even on difficult templates. These reagents are suitable for standard PCR applications, screening and high-throughput testing.
This PCR master mix is composed of Taq DNA polymerase and an optimized buffer system, including dNTPs and magnesium chloride (1.5 mM).
Long term storage at -20 °C. Product expiry at -20 °C is stated on the label. Optional: Store at +4 °C for up to 6 months.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.