ContactUsLinkComponent
LED Lighted Rimless 2X Magnifier
Catalogue # 470347-526
Fournisseur:  CARSON OPTICAL
undefined
LED Lighted Rimless 2X Magnifier
Catalogue # 470347-526
Fournisseur:  CARSON OPTICAL
Supplier Number:  RM-95
Spécifications
  • Description:
    LED lighted rimless 2X magnifier
  • Cat. no.:
    470347-526
Spécifications
À propos de cet article
This 2X lighted 3.5" rimless acrylic round magnifier with soft pouch, is lightweight and ergonomically shaped.
  • Built-in LED
  • Distortion-free viewing
  • Includes batteries and soft pouch
Uses three, included, G10 batteries (389/LR1130).