PYREX® Separating Funnels, Squibb, Glass, Corning
PYREX® Separating Funnels, Squibb, Glass, Corning
Supplier:   Corning
PYREX® Separating Funnels, Squibb, Glass, Corning
Supplier:  Corning
Certificates
PYREX® Separating Funnels, Squibb, Glass, Corning
PYREX® Separating Funnels, Squibb, Glass, Corning
8 Options Available Below
Product Details & Documents
Borosilicate glass funnels feature an unbreakable PTFE valve and drip tip and have a screw thread closure at the top. Supplied with a PTFE valve assembly and GL screw cap with PTFE-faced silicone liner. No stopper.
Replacement funnels do not include a PTFE valve and drip tip or cap.
Ordering information: For replacement valve bottoms or caps, contact your Avantor representative.
Product Family Options
Funnels, with bottom valve and cap
Informations sur le produit
  • Désignation
    Capacité
    Thread Finish
    Availability
    Price
  • Separatory Funnel with PTFE Valve and Threaded Screw Cap
    60 ml
    GL 32
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $343.00
    $343.00
    Case of 1
       $343.00
    Supplier Number: 6305-60
  • Separatory Funnel with PTFE Valve and Threaded Screw Cap
    250 ml
    GL45
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $410.00
    $410.00
    Case of 1
       $410.00
    Supplier Number: 6305-250
  • Separatory Funnel with PTFE Valve and Threaded Screw Cap
    500 ml
    GL45
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $484.00
    $484.00
    Case of 1
       $484.00
    Supplier Number: 6305-500
  • Separatory Funnel with PTFE Valve and Threaded Screw Cap
    1000 ml
    GL45
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $585.00
    $585.00
    Case of 1
       $585.00
    Supplier Number: 6305-1L
  • Separatory Funnel with PTFE Valve and Threaded Screw Cap
    2000 ml
    GL45
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $865.00
    $865.00
    Case of 1
       $865.00
    Supplier Number: 6305-2L
  • Separatory Funnel with PTFE Valve and Threaded Screw Cap
    4000 ml
    GL45
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $1,350.00
    $1,350.00
    Case of 1
       $1,350.00
    Supplier Number: 6305-4L
Funnels, without bottom valve and cap
Informations sur le produit
  • Désignation
    Capacité
    Thread Finish
    Availability
    Price
  • Funnel Only
    60 ml
    GL 32
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $212.00
    $212.00
    Case of 1
       $212.00
    Supplier Number: 6305-60FO
  • Funnel Only
    500 ml
    GL45
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $351.00
    $351.00
    Case of 1
       $351.00
    Supplier Number: 6305-500FO
  • Funnel Only
    2000 ml
    GL45
    Discontinued
    Supplier Number: -
  • Funnel Only
    6000 ml
    GL45
    Discontinued
    Supplier Number: -
Accessories
Informations sur le produit
  • Catalog #
    Size
    Availability
    Price
  • PTFE Valve Bottoms
    PTFE Valve Bottoms
    62890-658
    6 mm
    Log in pour la disponibilité
    Case of 1 - $226.00
Informations sur le produit
  • Catalog #
    Size
    Availability
    Price
  • PTFE Valve Bottoms
    PTFE Valve Bottoms
    62890-660
    8 mm
    Log in pour la disponibilité
    Case of 1 - $226.00