PYREX® Funnel, Plain, 60° Angle, Long Stem, Corning
 :   Corning
PYREX® Funnel, Plain, 60° Angle, Long Stem, Corning
 :  Corning
PYREX® Funnel, Plain, 60° Angle, Long Stem, Corning
PYREX® Funnel, Plain, 60° Angle, Long Stem, Corning
PYREX®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With a beaded edge for strength. Cone angle 60°. These funnels are fabricated from blanks formed to a 60° angle, which permits accurate fitting of filter paper and thus reducing filtering time. In addition, few funnels have depressed inside flutings which further decrease filtering time.
  • Catalog No:
  • 30230-500
  • 30230-503
  • 30230-929
  • 30230-984
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
Product Family Options
  • Désignation
    Stem Length
    Stem O.D.
    Top I.D.
  • Long Stem Funnel
    150 mm
    8 mm
    65 mm
    Case of 12
    Case of 12 - $254.15
    $254.15
    Case of 12
       $254.15
     : 6140-65
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    30230-500
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • Long Stem Funnel
    150 mm
    8 mm
    75 mm
    Case of 12
    Case of 12 - $262.65
    $262.65
    Case of 12
       $262.65
     : 6140-75
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    30230-503
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • Long Stem Funnel
    150 mm
    8 mm
    50 mm
    Case of 12
    Case of 12 - $328.00
    $328.00
    Case of 12
       $328.00
     : 6140-50
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    30230-929
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • Long Stem Funnel
    150 mm
    9 mm
    100 mm
    Case of 6
    Case of 6 - $187.15
    $187.15
    Case of 6
       $187.15
     : 6140-100
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    30230-984
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact