AE2000 Inverted Microscopes LED, Motic | Avantor
AE2000 Inverted Microscopes LED, Motic
Supplier:   Motic
AE2000 Inverted Microscopes LED, Motic
Supplier:  Motic
Certificates
AE2000 Inverted Microscopes LED, Motic
AE2000 Inverted Microscopes LED, Motic
4 Options Available Below
Product Details & Documents
The AE2000 is a revolutionary design that is packed with features and sets a new benchmark in affordable inverted microscopy. Motic’s AE2000 inverted microscope is designed for routine live cell inspection with an easy and flexible set up, ideal for university, routine-clinical, and lab environments.
  • Head Siedentopf type 45° inclined
  • Side facing quadruple nosepiece
  • CCIS Plan Achromat objective PL PH4×, PL PH 10× (basic package)
  • Optional CCIS Plan Achromat objective with PL PH 20×
  • ELWD N.A. 0.30 condenser
  • Phase slider Ph0, BDF, Ph1
  • Centering telescope
  • Plain stage with metal and glass stage inserts
  • 3 W LED illumination with intensity control
Binocular inverted live cell microscope for routine microbiology. The pre-defined optical setup with LWD CCIS© Phase objectives is ready for the inspection of diverse native samples in cell culture flasks. Lab technicians benefit from the convincing phase contrast concept, while the Fixed-Köhler illumination with interchangeable light source (HAL/LED) and auto ON/OFF function enables a fast throughput of samples.
Ordering information: Contact your Avantor representative to learn more about this and other Motic solutions. All Motic products come with a one-year electrical and five-year mechanical warranty.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • AE2000 binocular inverted microscope LED, basic package
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,342.54
    $3,342.54
    Each
       $3,342.54
    La quantité minimale de commande est de 1
    Supplier Number: 1100103800222
  • AE2000 trinocular inverted microscope LED, basic package
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,947.28
    $3,947.28
    Each
       $3,947.28
    La quantité minimale de commande est de 1
    Supplier Number: 1100103800266
  • AE2000 binocular inverted microscope LED, with PL PH 20×
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,758.24
    $3,758.24
    Each
       $3,758.24
    La quantité minimale de commande est de 1
    Supplier Number: 1100103800268
  • AE2000 trinocular inverted microscope LED, with PL PH 20×
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,363.92
    $4,363.92
    Each
       $4,363.92
    La quantité minimale de commande est de 1
    Supplier Number: 1100103800267