BA410E Compound Microscopes, Motic | Avantor
BA410E Compound Microscopes, Motic
Supplier:   Motic
BA410E Compound Microscopes, Motic
Supplier:  Motic
Certificates
BA410E Compound Microscopes, Motic
BA410E Compound Microscopes, Motic
2 Options Available Below
Product Details & Documents
The BA410E is Motic's flagship model, and it displays the highest level of optical performance. The BA410E also allows a profound diagnosis for those in pathology, haematology and cytology who require the best colour fidelity. Increased numerical apertures provide an expanded resolution power for a more detailed specimen evaluation.
  • Head Siedentopf type 30° inclined, 360° rotating [trinocular: (light split = 20/80)]
  • Reversed sextuple nosepiece
  • Coaxial coarse and fine focusing adjustment
  • Built in low position rackless stage with right hand control
  • Achromat swing-out condenser N.A. 0.90 with iris diaphragm
Binocular or trinocular research microscope for diverse questions from life sciences, but also from material sciences. The standard EC-H plan achromat objectives can be replaced by fluorite and apochromatic lenses for maximum optical performance. Demanding users benefit from the system’s flexibility of the BA410E construction kit, ready to configure a personal microscope for individual requirements.
Delivery information: Contact your Avantor representative to learn more about this and other Motic solutions. All Motic products come with a one-year electrical and five-year mechanical warranty.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • BA410E binocular compound microscope
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,515.34
    $4,515.34
    Each
       $4,515.34
    La quantité minimale de commande est de 1
    Supplier Number: 1100100404301
  • BA410E trinocular compound microscope
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,773.98
    $4,773.98
    Each
       $4,773.98
    La quantité minimale de commande est de 1
    Supplier Number: 1100100404311