AE31E LED Inverted Microscopes, Motic
 :   Motic
AE31E LED Inverted Microscopes, Motic
 :  Motic
AE31E LED Inverted Microscopes, Motic
AE31E LED Inverted Microscopes, Motic
The AE31 Elite is a refined inverted microscope with an extended focus on practical aspects. In order to harmonize the optical setup of our CCIS© infinity concept, the AE31 Elite works with the newest generation of LWD plan achromatic lenses.
  • Siedentopf binocular tube 45° inclined [trinocular: (light split: 20/80)]
  • Side facing quintuple nosepiece
  • ELWD N.A. 0.30 (W.D. 72 mm) condenser
  • Phase slider Ph0, BF, Ph1
  • Phase centering telescope
Binocular or Trinocular live cell microscope platform for advanced microbiology. The powerful LWD CCIS© phase objectives are designed for the inspection of native cell cultures. Lab technicians save time by benefitting from a clever illumination concept. The instrument can be upgraded with fuorescence for research tasks, while the 'Light memory' function and the auto ON/OFF mode enables a fast and safe throughput of samples.
 : Contact your Avantor representative to learn more about this and other motic solutions. All motic products come with a one-year electrical and five-year mechanical warranty.
Product Family Options
  • Désignation
  • AE31E LED binocular inverted microscope
    Each
    Each - $4,870.85
    $4,870.85
    Each
       $4,870.85
     : 1100100200861
  • AE31E LED trinocular inverted microscope
    Each
    Each - $5,498.16
    $5,498.16
    Each
       $5,498.16
     : 1100100200841