PYREX® Volumetric Flasks, Class A, Micro, Hexagonal Base, with Stopper, Corning | Avantor
PYREX® Volumetric Flasks, Class A, Micro, Hexagonal Base, with Stopper, Corning
   Options durables disponibles
Supplier:   Corning
PYREX® Volumetric Flasks, Class A, Micro, Hexagonal Base, with Stopper, Corning
Supplier:  Corning
   Options durables disponibles
Certificates
PYREX® Volumetric Flasks, Class A, Micro, Hexagonal Base, with Stopper, Corning
PYREX® Volumetric Flasks, Class A, Micro, Hexagonal Base, with Stopper, Corning
5 Options Available Below
Product Details & Documents
Flat-bottom, long neck flasks are for microchemical work
  • Hexagonal base improves stability
  • White markings are sharp and easy-to-read
  • Supplied with interchangeable [ST] hollow glass stoppers
Flask is designed in accordance with the recommendations of the committee of microchemical apparatus analytical division of the ACS. All flasks are graduated to class A volumetric tolerances and calibrated to meet ASTM specifications E-237. Certified flasks are individually serialized and supplied with NIST-traceable certificate of identification and capacity.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 29502-023
  • 89091-302
  • 29502-103
  • 89091-114
  • 89090-826
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
Spécifications
Product Family Options
Informations sur le produit
  • Capacité
    ST Stopper No.
    O.D.×H
    Tolerance
    Availability
    Price
  • 1 ml
    8
    8×68 mm (5/16×211/16")
    ±0.01 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 12
    Case of 12 - $1,300.00
    $1,300.00
    Case of 12
       $1,300.00
    Supplier Number: 5630-1
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    29502-023
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 2 ml
    8
    12×71 mm (1/2×213/16")
    ±0.15 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 12
    Case of 12 - $1,400.00
    $1,400.00
    Case of 12
       $1,400.00
    Supplier Number: 5630-2
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    89091-302
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 5 ml
    8
    17×80 mm (11/16×35/16")
    ±0.02 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 12
    Case of 12 - $1,340.00
    $1,340.00
    Case of 12
       $1,340.00
    Supplier Number: 5630-5
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    29502-103
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 10 ml
    9
    20×105 mm (13/16×41/8")
    ±0.02 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 12
    Case of 12 - $1,540.00
    $1,540.00
    Case of 12
       $1,540.00
    Supplier Number: 5630-10
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    89091-114
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 25 ml
    9
    29×132 mm (11/8×53/16")
    ±0.03 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 6
    Pack of 6 - $897.00
    $897.00
    Log in pour la disponibilité
    Case of 12
    Case of 12 - $1,794.00
    $1,794.00
    Pack of 6
       $897.00
    Case of 12
       $1,794.00
    Supplier Number: 5630-25
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    89090-826
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact