Eisco Schellbach Automatic Zero Glass Burettes with Two Stopcocks
À propos de cet article
The most accurate burettes available.
- Automatic zero burette mounted on a glass bottle reservoir via ground glass joint
- Designed to automatically set to zero mark upon pressure release
- White stripe with blue ribbon opposite graduations improves visibility/readability of the meniscus
- Two PTFE stopcocks - one for titration and one for drainage of excess liquid back into the reservoir
- Both burette and reservoir are made of heavy-duty borosilicate 3.3 glass
- White enamel graduations for contrast against dark liquids
- Class A and Class B versions available
Schellbach Automatic Zero Burettes are each mounted on a graduated reservoir bottle. Solution rises to the top of burette by squeezing the rubber bulb. The burette is automatically set to zero releasing the air pressure from the valve. Unique features of Schellbach design include a vertical white stripe with central blue ribbon to facilitate accurate reading of the meniscus, and an intermediate PTFE stopcock which facilitates the complete drainage of excess liquid back into the reservoir. Both burette and reservoir are made of precisely-bore, high quality, heavy-duty borosilicate 3.3 glass with excellent durability and chemical resistance. Both Class A and Class B versions conform with DIN ISO 385 standards for accuracy. Calibrated to deliver. All components are packed in custom foam inserts for safe transport and storage.
8 Options Available Below
Product Details & Documents
The most accurate burettes available.
- Automatic zero burette mounted on a glass bottle reservoir via ground glass joint
- Designed to automatically set to zero mark upon pressure release
- White stripe with blue ribbon opposite graduations improves visibility/readability of the meniscus
- Two PTFE stopcocks - one for titration and one for drainage of excess liquid back into the reservoir
- Both burette and reservoir are made of heavy-duty borosilicate 3.3 glass
- White enamel graduations for contrast against dark liquids
- Class A and Class B versions available
Schellbach Automatic Zero Burettes are each mounted on a graduated reservoir bottle. Solution rises to the top of burette by squeezing the rubber bulb. The burette is automatically set to zero releasing the air pressure from the valve. Unique features of Schellbach design include a vertical white stripe with central blue ribbon to facilitate accurate reading of the meniscus, and an intermediate PTFE stopcock which facilitates the complete drainage of excess liquid back into the reservoir. Both burette and reservoir are made of precisely-bore, high quality, heavy-duty borosilicate 3.3 glass with excellent durability and chemical resistance. Both Class A and Class B versions conform with DIN ISO 385 standards for accuracy. Calibrated to deliver. All components are packed in custom foam inserts for safe transport and storage.
Product Family Options
Class A
Informations sur le produit
- CapacitéGraduationJoint SizeReservoir CapacityToleranceAvailabilityPrice
- Supplier Number: CH0242P
- Supplier Number: CH0242Q
- Supplier Number: CH0242R
- Supplier Number: CH0242S
Class B
Informations sur le produit
- CapacitéGraduationJoint SizeReservoir CapacityToleranceAvailabilityPrice
- Supplier Number: CH0242T
- Supplier Number: CH0242U
- Supplier Number: CH0242V
- Supplier Number: CH0242W