CiDehol® 70 Pre-Saturated Cleanroom Wipes, Decon Labs
Catalog # 76579-806
Supplier: Decon Labs
Specifications
- Description:Cleaning wipes
- Cat. no.:76579-806
- Packaging:175/canister
- W×L:5×8"
Specifications
About this item
CiDehol® 70 wipes are pre-saturated with 70% isopropyl alcohol.
- Tear-resistant
- Lint-free
- Unique self-closing lid keeps wipes saturated
CiDehol® 70 wipes are strong, lint-free wipes made for use on hard surfaces in cleanrooms, labs, and other areas where a pre-saturated cleaning wipe is a convenience. Wipes are 100% PET, non-woven. Each wipe measures 5×8". Packaged 175 wipes per canister.