The Wizard® Genomic Plant DNA Purification Kit
Fournisseur: PROMEGA SUNNYVALE INC SE
Certificats
À propos de cet article
A simple method to isolate genomic DNA from plants.
- Extract plant DNA with a simple, 60-minute scalable protocol
The Wizard® Genomic DNA Purification Kit provides a simple, solution-based method for isolation of DNA. DNA purified with this system is suitable for a variety of applications, including amplification, digestion with restriction endonucleases and membrane hybridizations (e.g., Southern and dot/slot blots).
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together