VWR® TraceClean® Low TOC Vials | Avantor
VWR® TraceClean® Low TOC Vials
VWR® TraceClean® Low TOC Vials
Supplier:  VWR International
Certificates
VWR® TraceClean® Low TOC Vials
2 Options Available Below
Product Details & Documents
These glass vial are used primarily for the collection and testing of aqueous samples for Total Organic Carbon (TOC). Secondary uses include containment of extracts. Please refer to the applicable certificate of analysis for certified analytes and corresponding reporting limits.
  • Certificate of analysis and cleaning protocol form included in each product case
  • Cast PTFE thin septa cap with dust covers
For pre-cleaned and certified products, a lot certificate of analysis is generated with the analytical results obtained including the concentration reporting limits and units of measurement.
D10 protocol is a depyrognation-like process for low level Total Organic Carbon (TOC). Caps are 1-piece Open top polypropylene with cast PTFE septa ultrasonically bonded (not glued); overall thickness 0.060 ±0.005" (silicone= 0.056", PTFE= 0.004"). The PTFE liner is a 0.10" thick disc.
Certifications: Meets ASTM standards. EN 342.
Documents
Product Family Options
Informations sur le produit
  • Désignation
    Capacité
    Diameter ×H
    Color
    Avec bouchon
    Cap Type
    Taille du bouchon
    Cap Color
    Septum
    Availability
    Price
  • TOC vials, <10 ppb TOC
    40.00 ml
    27.5×95 mm
    Clear
    Oui
    Open top, 1-piece
    24-414
    White
    Cast thin PTFE
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 80
    Case of 80 - $352.24
    $352.24
    Prix promotionnel pour les nouveaux clients
    Case of 80
       $352.24
  • TOC vials, <20 ppb TOC
    40.00 ml
    27.5×95 mm
    Clear
    Oui
    Open top, 1-piece
    24-414
    White
    Cast thin PTFE
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 80
    Case of 80 - $265.99
    $265.99
    Prix promotionnel pour les nouveaux clients
    Case of 80
       $265.99