ContactUsLinkComponent
Native barcoding auxiliary kit v14, Oxford Nanopore Technologies
Catalogue # 76595-412
Numéro CAS:  
undefined
Native barcoding auxiliary kit v14, Oxford Nanopore Technologies
Catalogue # 76595-412
Supplier Number:  EXP-NBA114
Numéro CAS:  
Please note that Oxford Nanopore Technologies requires the acceptance of their Terms & Conditions prior to product order placement. If you select product(s) to order, the Terms and Conditions will be displayed in the shopping cart for your review and acceptance.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Native barcoding auxiliary kit v14
  • Compatible with:
    R10.4.1 flow cells and kit 14 chemistry
  • Cat. no.:
    76595-412
  • No. of Reactions:
    12
Spécifications
À propos de cet article
This expansion is recommended for users who want to make use of unused barcodes to maximise the use of their Native Barcoding Kits.
This expansion pack contains additional library preparation reagents for the Native Barcoding Kit 24 and 96 V14 (SQK-NBD114.24 and SQK-NBD114.96) for another 12 reactions in conjunction with the Sequencing Auxiliary Vials V14 (EXP-AUX003).
The Native Barcoding Auxiliary Kit V14 (EXP-NBA114) can only be used with Kit 14 chemistry. This kit is not compatible with previous kits chemistries.
Ordering information: Kits are shipped at 2 to 8 °C in an environmentally-friendly temperature-controlled shipping box. Upon receipt, please place the product in a long-term storage location at ‒20 °C. Oxford Nanopore Technologies deem the useful life of the product to be 3 months from receipt by the customer.
Caution: Oxford Nanopore Technologies products are not intended for use for health assessment or to diagnose, treat, mitigate, cure, or prevent any disease or condition.