Heated Forceps, Mopec
Heated Forceps, Mopec
Marketsource
Supplier:   Mopec
Heated Forceps, Mopec
Supplier:  Mopec
Marketsource
Certificates
Heated Forceps, Mopec
Heated Forceps, Mopec
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Mopec’s heated forceps are electrically heated instruments designed to assist with the proper positioning of tissue specimens during the embedding process in paraffin wax. The safe, low-voltage heating element maintains a steady temperature throughout specimen manipulation.
  • Designed to take up minimal bench-top space
  • Anti-microbial coating to help prevent bacteria and virus growth
  • Molded finger grips increase user comfort and remain cool when in use
  • Bayonet connection prevents disconnection while in use
The ergonomic design ensures user comfort while performing specimen placement, and the anti-microbial coating helps prevent bacteria and virus growth. Choose between serrated or smooth tips. Available in three sizes and compatible with the Mopec control unit.
Product Family Options
Product Information
  • Désignation
    Color
    Tip Style
    Tip Width
    Availability
    Price
  • Heated forceps
    Red
    Serrated
    1 mm
    Log in for availability
    Log in for availability
    Each
    Each - $1,102.50
    $1,102.50
    Each
       $1,102.50
    Supplier Number: BK301-SE
  • Heated forceps
    Red
    Smooth
    1 mm
    Log in for availability
    Log in for availability
    Each
    Each - $1,102.50
    $1,102.50
    Each
       $1,102.50
    Supplier Number: BK301-SM
  • Heated forceps
    Yellow
    Serrated
    2 mm
    Log in for availability
    Log in for availability
    Each
    Each - $1,102.50
    $1,102.50
    Each
       $1,102.50
    Supplier Number: BK302-SE
  • Heated forceps
    Yellow
    Smooth
    2 mm
    Discontinued
    Supplier Number: BK302-SM
    Specifications:
    Marketsource
    Marketsource
  • Heated forceps
    Blue
    Serrated
    4 mm
    Log in for availability
    Log in for availability
    Each
    Each - $1,100.04
    $1,100.04
    Each
       $1,100.04
    Supplier Number: BK304-SE
  • Heated forceps
    Blue
    Smooth
    4 mm
    Discontinued
    Supplier Number: BK304-SM
    Specifications:
    Marketsource
    Marketsource