Biohazard Waste Bag and Bin, Mopec
Catalog # 76579-394
Supplier: Mortech Manufacturing
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Biohazard waste bags
- Cat. no.:76579-394
Specifications
About this item
Mopec’s biohazard waste bags are made of a durable, autoclavable, 2.25 HDPE material and are clearly marked with the 'Biohazard' symbol.
- Made from a durable, autoclavable, 2.25 HDPE material
- Dimensions: 25×35"
- Capacity: 12 to 16 galloons
- Can withstand temperatures up to 250 ˚F during autoclaving
- Feature OSHA-standard 'Biohazard' symbol and words 'Hazardous Waste'
- Color-changing sterilization indicator
- Features large, white marking panel for use with permanent markers
- Fits perfectly into Biohazard Waste Bin (BG681-B, sold separately)
Bags feature drawstring loops, these bags allow for proper ventilation during sterilization and provide the user with a safe and easy way to remove bags from a hot autoclave. Capacity of 12 to 16 gallons. Sold by the case of 100 bags.