Formaway Formalin Neutralizing Absorption Pads, Mopec | Avantor
Formaway Formalin Neutralizing Absorption Pads, Mopec
Formaway formalin neutralizing absorption pads, 3×4"
Catalog # 76579-376
Supplier:   Mopec
Fournisseur #:  BE092
Formaway Formalin Neutralizing Absorption Pads, Mopec
Catalog # 76579-376
Supplier:  Mopec
CAS Number:  
Formaway Formalin Neutralizing Absorption Pads, Mopec
Formaway Formalin Neutralizing Absorption Pads, Mopec
Formaway formalin neutralizing absorption pads, 3×4"
CAS Number:  
Price
$493.52
Log in pour la disponibilité
Pack of 100 (1/Itm/Bx)
Product Details & Documents
Mopec’s Formaway Formalin Neutralizing Absorbent Pad (F-NAP) was designed to reduce formaldehyde vapors while dissecting tissue, transferring tissue from containers of formalin, and transporting or storing tissue in containers of formalin, by absorbing and neutralizing formalin.
  • F-NAP provides excellent vapor control during gross examination, dissecting, transporting, or transferring formalin fixed tissue
  • F-NAP Pads are also used for vapor control in storage of formalin, formalin fixed tissue containers and for small formalin spills
  • Blue color allows for easier viewing of small specimens, and the soft cutting surface allows for longer lasting scalpels
  • Sold in packs of 100
Mopec’s Formaway formalin neutralizing absorbent pad (F-NAP) was designed to facilitate the reduction of formaldehyde vapors by absorbing and neutralizing formaldehyde, glutaraldehyde and other aldehydes. Once the formalin solution is absorbed, the formalin neutralizing absorbent pad will convert formaldehyde and other aldehydes to a non-hazardous polymer, eliminating the source of harmful vapors making the work area safer.
Spécifications
Spécifications
  • Désignation:
    Formaway formalin neutralizing absorption pads
  • Taille:
    3×4"